Learn More
Buffer solution, pH 6.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
26.40€ - 76.40€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346089
|
Thermo Scientific Alfa Aesar
042418.AP |
500 mL |
26.40€
500mL |
In Stock
Log in to see stock. |
|||||
11374578
|
Thermo Scientific Alfa Aesar
042418.K2 |
1 L |
41.60€
1 L |
In Stock
Log in to see stock. |
|||||
11364578
|
Thermo Scientific Alfa Aesar
042418.K7 |
4 L |
76.40€
4 L |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
Buffer solution (pH 6) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.