Learn More
Budesonide-d6, TRC
Product Code. 30335969
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335969 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335969 Supplier Toronto Research Chemicals Supplier No. TRCB6894931MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Budesonide-d6
CAS 1134189-63-1
Molecular Formula C25H28D6O6
Quantity 1 mg
Synonym (11β,16α)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione-d6, Bidien-d6, Budenofalk-d6, Budesomide-d6, Budeson-d6, Cortivent-d6, Entocort-d6, Micronyl-d6, Nebuampul-d6, Preferid-d6, Pulmicort-d6, Respules-d6, Rhinocort-d6, Rhinocort Aqua-d6-d6, S 1320-d6, Spirocort-d6, Turbuhaler-d6
InChI Formula InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3
SMILES O=C1C([2H])=C[C@@]2(C)C(C([2H])([2H])C[C@]3([H])[C@]2([H])[C@H](O)C[C@@]4(C)[C@]3([H])C[C@@H](OC(CCC)O5)[C@]45C(C([2H])([2H])O)=O)=C1[2H]
IUPAC Name 11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
Molecular Weight (g/mol) 436.57
Formula Weight 436.2732