Learn More
BUCHI Reitmeyer Adapters for Rotavapor
Shop All Buchi Products
Click to view available options
Connection Type:
STJ 29/32
Unit Size:
Each
Description
- Reitmeyer style
- Made from glass
- SJ29/32
- Length: 135 mm
Specifications
Specifications
| Connection Type | STJ 29/32 |
| Description | Reitmeyer adapter |
| Material | Borosilicate Glass (3.3) |
| Type | Rotary Evaporator Adapter |
| For Use With (Application) | Prevents contents from the evaporation flask being drawn into the condenser in the event of bumping |