Learn More
BUCHI Receiving Flasks for Rotary Evaporators
Product Code. p-3649089 Shop All Buchi Products
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
3000 mL
Material (Coating):
P+G
P+G LT
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Material (Coating) Unit Size
10416011 500 mL P+G Each
10489492 1000 mL P+G Each
12792455 2000 mL P+G LT Each
10027151 3000 mL P+G Each
10601891 500 mL P+G LT Each
12786174 3000 mL P+G LT Each
12766174 1000 mL P+G LT Each
10392272 2000 mL P+G Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10416011 Supplier BUCHI Supplier No. 025264
Add to basket
Add to basket
Use these receiving flasks with any rotary evaporator. BUCHI Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
  • Spherical Receiving Flasks for rotary evaporators with ball joint (35/20)
  • Exclusive use of DURAN borosilicate glass 3.3
  • High chemical resistance against acids, alkalis and organic substances
  • Resistant to thermal shocks and high temperatures combined with low thermal expansion
  • Available in volumes from 50 mL to 3000 mL
  • Available uncoated, with P+G coating or P+G LT for dry ice condensers
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Receiving Flask
For Use With (Equipment) Rotary evaporator
Includes Ball joint KS-35
Material (Coating) P+G
Shape Round
Taper 35/20
Volume (Metric) 500 mL