Learn More
BUCHI Receiving Flasks for Rotary Evaporators
Shop All Buchi Products
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
3000 mL
Material (Coating):
P+G
P+G LT
Unit Size:
Each
Description
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
- Spherical Receiving Flasks for rotary evaporators with ball joint (35/20)
- Exclusive use of DURAN borosilicate glass 3.3
- High chemical resistance against acids, alkalis and organic substances
- Resistant to thermal shocks and high temperatures combined with low thermal expansion
- Available in volumes from 50 mL to 3000 mL
- Available uncoated, with P+G coating or P+G LT for dry ice condensers
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Type | Receiving Flask |
| For Use With (Equipment) | Rotary evaporator |
| Includes | Ball joint KS-35 |
| Material (Coating) | P+G LT |
| Shape | Round |
| Volume (Metric) | 3000 mL |