Learn More
BUCHI Deuterium Lamp for Chromatography
Shop All Buchi Products
Click to view available options
Product Type:
Deuterium Lamp
Unit Size:
Each
Description
- Used in preparative chromatography
- Intensity Range: 190 to 740nm
Specifications
Specifications
| Light Source | Deuterium |
| Wavelength | 740/190 nm |
| For Use With (Equipment) | UV photometer C-635/630 in preparative chromatography |
| Product Type | Deuterium Lamp |