Learn More
BUCHI Condensers for BUCHI Rotavapors
Product Code. 10215222 Shop All Buchi Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-210/R-215
Rotavapor R-210/R-215 R-210/R-215
Material (Coating):
None
P+G
Type:
Condenser C
Condenser S
Diagonal Condenser
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Material (Coating) Type Unit Size
10215222 Rotavapor R-210/R-215 None Condenser C Each
11977198 Rotavapor R-210/R-215 R-210/R-215 P+G Diagonal Condenser Each
10077051 Rotavapor R-210/R-215 None Condenser S Each
10302282 Rotavapor R-210/R-215 P+G Condenser S Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10215222 Supplier BUCHI Supplier No. 040640
Estimated Shipment: 14-01-2027
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Condenser C
For Use With (Application) Evaporator Condensation
For Use With (Equipment) Rotavapor R-210/R-215
Material (Coating) None
Quantity 1 Ea.