Learn More
BUCHI Condensers for BUCHI Rotavapors
Shop All Buchi Products
Click to view available options
For Use With (Equipment):
Rotavapor R-210/R-215
Rotavapor R-210/R-215 R-210/R-215
Material (Coating):
None
P+G
Type:
Condenser C
Condenser S
Diagonal Condenser
Unit Size:
Each
Description
- High throughput due to advanced design of condensers
- Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
- Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
- Maximum stability and safety with plastic and glass coating
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Type | Condenser S |
| For Use With (Application) | Evaporator Condensation |
| For Use With (Equipment) | Rotavapor R-210/R-215 |
| Material (Coating) | None |
| Quantity | 1 Ea. |