Learn More
Paragon Scientific™ BS/IP/SL Viscometer for Transparent Liquids (non certified)
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
| Viscosity Range | 2000 to 10,000 cSt |
| Calibrated | No |
| Approximate Constant | 10 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | 4 |
4 week lead-time