missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bryostatin 1
GREENER_CHOICE

Product Code. 18084822
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084822 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18084822 Supplier Tocris Bioscience™ Supplier No. 2383/10U
  • ONLINE EXCLUSIVE PRICE
    418.00€
    394.80€ / 10µg
    Save 23.20€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Protein kinase C activator

Protein kinase C (PKC) activator that binds with high affinity (Ki = 1.35 nM). Initially activates and subsequently induces downregulation of PKC isozymes. Sensitizes tumor cells to cytotoxic effects of anticancer agents. Restores hippocampal synapses and spatial learning and memory in an in vivo model of fragile X syndrome.
TRUSTED_SUSTAINABILITY

Specifications

CAS 83314-01-6
Quantity 10 μg
Molecular Formula C47H68O17
Formula Weight 905.04
Purity >98%
Molecular Weight (g/mol) 905.044
Chemical Name or Material Bryostatin 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.