Learn More
Steris™ Browne™ Control™ Autoclave Indicator Valves
Click to view available options
Color:
Black Spot
Green Spot
White Spot
Yellow Spot
missing translation for 'unitSize'
Pack of 100
Beschreibung
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
- The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
- Various colors
- Glass tubes
Spezifikation
Spezifikation
| Color | White Spot |
| For Use With (Application) | For monitoring porous load steam sterilization processes |
| For Use With (Equipment) | Autoclave |
| Product Type | Browne Control Tube |
| Quantity | 100 Pk. |
| Sterilization Indicator | Yes |
| Sterilization Method | Steam, Dry Heat |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur