Learn More
Hirschmann™ Class A Amber Glass Volumetric Flasks with Poly Stopper
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
7/16 NS Standard Ground
Tolerance:
±0.02 mL
±0.025 mL
±0.03 mL
±0.04 mL
±0.06 mL
±0.08 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
missing translation for 'unitSize'
Each
Pack of 2
Description
- With NS ground and polyethylene stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Spécification
Spécification
| Certifications/Compliance | USP |
| Closure Material | Polyethylene |
| Color | Brown |
| Capacity (Metric) | 5 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 70 mm |
| Limit of Error | ±0.02 mL |
| Standard Taper Joint | 10/19 NS Standard Ground |
| Tolerance | ±0.02 mL |
| Graduated | Yes |
| Afficher plus |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur