Learn More
Brompheniramine Maleate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Brompheniramine maleate |
| CAS | 980-71-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H19 Br N2 . C4 H4 O4 |
| Quantity | 1 g |
| Synonym | 2-Pyridinepropanamine, γ-(4-bromophenyl)-N,N-dimethyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[p-bromo-α-[2-(dimethylamino)ethyl]benzyl]-, maleate (1:1) (8CI), (±)-Brompheniramine maleate, 2-[p-Bromo-α-(2-dimethylaminoethyl)benzyl]pyridine maleate, 2-[p-Bromo-α-[2-(dimethylamino)ethyl]benzyl]pyridine bimaleate, Allent, Bromopheniramine maleate, Brompheniramine hydrogen maleate, Brompheniramine maleate, Dimetane, Dimetane maleate, Dimotane, Ilvin, Ilvin maleate, Lodrane, Nagemid, Parabromdylamine maleate, Parabromodylamine maleate, Symptom 3, Veltane, dl-Bromopheniramine maleate, dl-Brompheniramine maleate |
| InChI Formula | InChI=1S/C16H19BrN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CN(C)CCC(c1ccc(Br)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | 3-(4-bromophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine;(Z)-but-2-enedioic acid |
| Molecular Weight (g/mol) | 435.31 |
| Show More |