Learn More
Bromotripyrrolidinophosphonium Hexafluorophosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Bromotripyrrolidinophosphonium Hexafluorophosphate |
| CAS | 132705-51-2 |
| Molecular Formula | C12H24BrF6N3P2 |
| Quantity | 25 g |
| Synonym | (T-4)-Bromotri-1-pyrrolidinyl-phosphorus(1+) Hexafluorophosphate(1-), Pyrrolidine Phosphorus Complex, Bromotris(pyrrolidino)phosphonium Hexafluorophosphate |
| InChI Formula | InChI=1S/C12H24BrN3P.F6P/c13-17(14-7-1-2-8-14,15-9-3-4-10-15)16-11-5-6-12-16;1-7(2,3,4,5)6/h1-12H2;/q+1;-1 |
| SMILES | Br[P+](N1CCCC1)(N2CCCC2)N3CCCC3.F[P-](F)(F)(F)(F)F |
| IUPAC Name | bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)salt |
| Molecular Weight (g/mol) | 466.18 |
| Formula Weight | 465.05 |