Learn More
(Bromomethyl-d2)cyclopropane-1-d1, CDN
Product Code. 30317717
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317717
Brand: CDN CDND59820.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(Bromo-dideuteriomethyl)-1-deuteriocyclopropane
CAS 1219799-17-3
Molecular Formula C4 2H3 H4 Br
Quantity 0.25 g
Synonym 1-(Bromo-dideuteriomethyl)-1-deuteriocyclopropane, (Bromomethyl-d2)cyclopropane-1-d1
InChI Formula InChI=1 S/C4H7Br/c5-3-4-1-2-4/h4H,1-3H2/i3D2,4 D
SMILES [2 H]C([2 H])(Br)C1([2 H])CC1
IUPAC Name 1-[bromo(dideuterio)methyl]-1-deuteriocyclopropane
Molecular Weight (g/mol) 138.02
Formula Weight 136.9919 g/mol
Percent Purity 98 atom % D, min 97% Chemical Purity