Learn More
Bromoethane-d5, TRC
Product Code. 30331286
Click to view available options
Quantity:
1 g
Unit Size:
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30331286
Brand: Toronto Research Chemicals TRCB6841971G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bromoethane-D5
CAS 3675-63-6
Purity Grade Notes GC
Molecular Formula C2 2H5 Br
Quantity 1 g
Synonym Ethane-1,1,1,2,2-d5, 2-bromo- (ACI), 2-Bromoethane-1,1,1,2,2-d5 (ACI), Ethane-d5, bromo- (6CI, 7CI, 8CI, 9CI), 1-Bromo-1,1,2,2,2-pentadeuterioethane, Bromoethane-d5, Ethyl bromide-d5, Ethyl-d5 bromide, Pentadeuterioethyl bromide, Pentadeuteroethyl bromide, Perdeuterioethyl bromide, Bromoethane-D5
InChI Formula InChI=1S/C2H5Br/c1-2-3/h2H2,1H3/i1D3,2D2
SMILES [2H]C([2H])([2H])C([2H])([2H])Br
IUPAC Name 1-bromo-1,1,2,2,2-pentadeuterioethane
Molecular Weight (g/mol) 114
Formula Weight 112.9888
Percent Purity >95