Learn More
Bromobenzene-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Bromobenzene-2,3,5,6-D4 |
| CAS | 64646-03-3 |
| Molecular Formula | C6 D4 H Br |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,4,5-d4, 3-bromo- (9 CI), Bromobenzene-2,3,5,6-d4, Bromobenzene-2,3,5,6-D4, Bromobenzene-d4, 3-Bromobenzene-1,2,4,5-d4 |
| InChI Formula | InChI=1 S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5 H/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1cc([2 H])c([2 H])c(Br)c1[2 H] |
| IUPAC Name | 3-bromo-1,2,4,5-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 161.0326 |
| Formula Weight | 159.9826 g/mol |
| Show More |