missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromo 2,3,4,6-Tetra-O-acetyl-Alpha-D-galactopyranoside(Stabilized with 2% (wt/wt) CaCO3), TRC
Specifications
Specifications
| Chemical Name or Material | Bromo 2,3,4,6-Tetra-O-acetyl-a-D-galactopyranoside |
| CAS | 3068-32-4 |
| Molecular Formula | C14 H19 Br O9 |
| Quantity | 10 g |
| Synonym | α-D-Galactopyranosyl bromide, 2,3,4,6-tetraacetate, Galactopyranosyl bromide, tetraacetate, α-D- (6CI,7CI,8CI), α-D-Galactopyranosyl bromide, tetraacetate (9CI), (2R,3S,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, 1-Bromo-2,3,4,6-tetra-O-Acetyl-α-D-galactopyranose, 1-α-Bromo-2,3,4,6-tetraacetyl-D-galactopyranose, 2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide, 2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl-1-bromide, 2,3,4,6-Tetra-O-acetyl-α-D-galactosyl bromide, 2,3,4,6-Tetraacetyl-α-D-galactopyranosyl bromide, 2,3,4,6-Tetraacetyl-α-D-galactosyl bromide, Acetobromo-α-D-galactose, Acetobromogalactose, Acetylbromogalactose, Tetra-O-acetyl-α-D-galactopyranosyl bromide, Tetra-O-acetyl-α-D-galactosyl bromide, Tetraacetyl-α-D-galactopyranosyl bromide, α-Acetobromogalactose, α-Bromotetraacetylgalactose, α-D-Acetobromogalactose |
| InChI Formula | InChI=1S/C14H19BrO9/c1-6(16)20-5-10-11(21-7(2)17)12(22-8(3)18)13(14(15)24-10)23-9(4)19/h10-14H,5H2,1-4H3/t10-,11+,12+,13-,14+/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | [(2R,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 411.2 |
| Formula Weight | 410.0212 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?