Learn More
Bromanilic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Bromanilic Acid |
| CAS | 4370-59-6 |
| Molecular Formula | C6H2Br2O4 |
| Quantity | 500 mg |
| Synonym | 2,5-Dibromo-3,6-dihydroxy-2,5-cyclohexadiene-1,4-dione, 2,5-Dibromo-3,6-dihydroxy-p-benzoquinone, 2,5-Dibromo-3,6-dihydroxyquinone, 2,5-Dihydroxy-3,6-dibromo-1,4-benzoquinone, NSC 36928 |
| InChI Formula | InChI=1S/C6H2Br2O4/c7-1-3(9)5(11)2(8)6(12)4(1)10/h9,12H |
| SMILES | OC1=C(Br)C(=O)C(=C(Br)C1=O)O |
| IUPAC Name | 2,5-dibromo-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 297.89 |
| Formula Weight | 295.832 |