Learn More
Kinematica Polytron™ Homogenizer Base Stand for PT 1200 E
Product Code. 10283722 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
Polytron PT 1200E Homogenizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10283722 Polytron PT 1200E Homogenizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10283722 Supplier Kinematica Supplier No. 11040006
  • ONLINE EXCLUSIVE PRICE
    221.55€
    203.00€ / Pack of 1
    Save 18.55€
    8% Off
Estimated Shipment: 22-12-2026
Holder for PT 1200 Homognezizer and 4 Generators.
Stainless steel holder for drive unit, tabletop power supply, four dispersing aggregates and two tools.
Specifications
Product Type Base Stand
Material Stainless Steel
For Use With (Equipment) Polytron PT 1200E Homogenizer