Learn More
Brilliant Yellow (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Brilliant Yellow (Technical Grade) |
| CAS | 3051-11-4 |
| Molecular Formula | C26H18N4Na2O8S2 |
| Quantity | 250 mg |
| Synonym | 2,2'-(1,2-ethenediyl)bis[5-[2-(4-hydroxyphenyl)diazenyl]-Benzenesulfonic acid sodium salt, 2,2’-(1,2-ethenediyl)bis[5-[(4-hydroxyphenyl)azo]-Benzenesulfonic acid disodium salt, Brilliant Yellow C, C.I. Direct Yellow 4, C.I. Direct Yellow 4, disodium salt, Benzo Brilliant Yellow, Brilliant Fast Yellow G2N, Brilliant Paper Yellow, Brilliant Paper Yellow C, Brilliant Paper Yellow P, Brilliant Yellow (indicator), C.I. 24890, Calcomine Brilliant Paper Yellow, Chlorazol Brilliant Yellow 3G, Diaphtamine Brilliant Fast Yellow 3G, Diazol Brilliant Yellow N, Direct Brilliant Yellow G, Direct Yellow 4, Fenamin Yellow 3G, Kayaku Direct Yellow G, Nippon Brilliant Yellow G, Paper Yellow 3GX, Paper Yellow 3GXA, Paper Yellow CB, Tertrodirect Yellow B, Trisulfon Paper Yellow |
| InChI Formula | InChI=1S/C26H20N4O8S2.2Na/c31-23-11-7-19(8-12-23)27-29-21-5-3-17(25(15-21)39(33,34)35)1-2-18-4-6-22(16-26(18)40(36,37)38)30-28-20-9-13-24(32)14-10-20;;/h1-16,31-32H,(H,33,34,35)(H,36,37,38);;/q;2*+1/p-2/b2-1+,29-27?,30-28?;; |
| SMILES | [Na+].[Na+].Oc1ccc(cc1)N=Nc2ccc(\C=C\c3ccc(cc3S(=O)(=O)[O-])N=Nc4ccc(O)cc4)c(c2)S(=O)(=O)[O-] |
| IUPAC Name | disodium;5-[(E)-(4-hydroxyphenyl)diazenyl]-2-[(E)-2-[4-[(E)-(4-hydroxyphenyl)diazenyl]-2-sulfonatophenyl]ethenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 624.55 |
| Formula Weight | 624.0361 |