Learn More
Thermo Scientific™ Brilliant Green Agar
Product Code. p-8394024
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Brilliant Green Agar (Mod.)
Fill Weight:
17.5g ± 5%
19.5g ± 1.0g
Unit Size:
Pack of 10
Product Code. 12992588
Brand: Thermo Scientific™ PO0171A
Select and identify salmonellae (other than Salmonella typhi) fromfood, feed, water, and clinical samples with this diagnostic prepared media.
Select and identify salmonellae (other than Salmonella typhi) from food, feed, water, and clinical samples with Thermo Scientific™ Brilliant Green Agar (Modified) diagnostic medium. The modified formula provides improved inhibition of Escherichia coli and Proteus species. The medium has been widely assessed in Europe and has been used in ISO standards.
Brilliant Green Agar (Modified) provides greater inhibition of Escherichia coli and Proteus species than many other media formulations for the isolation of salmonellae. Growth of Pseudomonas species, whose colonies may resemble salmonellae on standard Brilliant Green Agar, are restricted, reducing confusion or extra work to confirm their identity. Inhibition of small numbers of salmonellae is absent from the modified formula1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Color Green / Brown
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Media Type Brilliant Green Agar (Mod.)
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.