Learn More
MP Biomedicals™ Briggs N Salt Mixture, Powder
Product Code. 11405632 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405632 2 kg 2kg
11415632 10 kg 10kg
11425632 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11405632 Supplier MP Biomedicals™ Supplier No. 0290283402
  • ONLINE EXCLUSIVE PRICE
    204.00€
    194.25€ / 2kg
    Save 9.75€
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Salt mixture for research animal diet.
  • Salt Mixture
  • For research animal diet
  • Powder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
For Use With (Application) Research animal diet
Quantity 2 kg