Learn More
Brema Ice Makers™ GB902AX Model Flaked Ice Maker
Product Code. 11967567
Change view
Click to view available options
Depth (Metric):
690mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
11967567 690mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11967567 Supplier Brema Ice Makers Supplier No. GB902AHC
  • ONLINE EXCLUSIVE PRICE
    4169.47€
    3205.95€ / Pack of 1
    Save 963.52€
    23% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Use this self-contained under-counter granular ice flaker in the pharmaceutical industry and for general scientific lab applications. Brema Ice Makers™ GB902AX Model Flaked Ice Makers can be installed anywhere at low cost and is designed for quick and easy maintenance operations.
  • Granular ice: 80-82% dry
  • 1l (water): 1kg (ice)
  • AISI 304 SS Scotch Brite finish
  • Tropical class +43°C
  • Adjustable Stainless Steel feet
  • Removable and cleanable air filter
  • AISI 304 SS evaporator
  • Air cooling system
  • Easy cleanable internal rounded surfaces
  • HCFC-Free insulated storage
  • Dust-proof water basin
  • RoHS Free
  • Disappearing insulated door
  • Electromechanical operation
  • ON/OFF rocker switch
  • Automatic water charge
Specifications
Capacity (Metric) 20 kg
Depth (Metric) 690mm
Product Type Flaked Ice Maker
Height (Metric) 500mm
Width (Metric) 660mm
Yield 90kg /24h
Electrical Requirements 230V, 50Hz, 550w