Learn More
Brady™ Breaker Blocker Component - Green Blocking Bar
Product Code. 15643517 Shop All Brady Products
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15643517 Green Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15643517 Supplier Brady™ Supplier No. 090893
  • ONLINE EXCLUSIVE PRICE
    35.80€
    31.29€ / Pack of 5
    Save 4.51€
    12% Off
Estimated Shipment: 06-01-2027
Helps to prevent electrical risk.
  • Green bar indicates locked ‘on’
Specifications
Product Type Breaker Blocker Components
Alternate Part Number EZ 480/600 VOLT 7.6 in. GREEN BLOCK BAR PK5
Color Green
Description Breaker Blocker Components - 190 mm Green Blocking Bar
Quantity 5 Pk.