Learn More
Aalborg™ Precision Valve for Brass Flowmeter
Product Code. 11728268
Click to view available options
Airflow Rate:
1000 mL/min.
200 mL/min.
21500 mL/min.
2500 mL/min.
400 mL/min.
6200 mL/min.
Waterflow Rate:
12mL/min.
200mL/min.
26mL/min.
650mL/min.
6mL/min.
80mL/min.
Unit Size:
Each
Product Code. 11728268
Brand: Aalborg™ VM1BB
Only null left
Add to basket
Add to basket
Aalborg™ Precision Valve for Brass Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • For high precision
  • Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • They may also be used as shut off valves.
  • Bodies of the valves are reinforced by structurally rigid metallic shells.
  • Shells are made of brass, bushings are made of plated brass or 316 stainless steel.
Specifications
Airflow Rate 200 mL/min.
Waterflow Rate 6mL/min.
For Use With (Equipment) 150 mm Gas/Liquid Flowmeter
Type Standard Valve