Learn More
Brady™ Brass Body Padlock
Product Code. 15693527
Click to view available options
Width (Metric) Shackle:
24.5 mm
32.5 mm
42 mm
No. of Pins:
4
5
Unit Size:
Pack of 6
Product Code. 15693527
Brand: Brady™ 805832
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Corrosion resistant.
  • Contains precision pin tumbler system
Specifications
Product Type Brass Padlock
Color Brass
Material Steel
Description Brass Padlocks
Diameter (Metric) Shackle 6.5 mm
Dimensions (L x W) 39.00 x 66.00 mm
Height (Metric) 66 mm
Height (Metric) Shackle 23.5 mm
Material (Shackle) Hardened Steel
No. of Pins 5
Quantity 6 Pk.
Width (Metric) 39 mm
Width (Metric) Shackle 32.5 mm