Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beakers
Product Code. 15338644
Click to view available options
Capacity (English):
8.4 oz.
20.28 oz.
Capacity (Metric):
250 mL
600 mL
Material:
Glass
Stainless Steel
Unit Size:
Each
Product Code. 15338644
Brand: Branson Ultrasonics™ 000140001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Bransonic ultrasonic baths
  • Suitable for all models
  • Available in glass, stainless steel, or polypropylene
Specifications
Type Beaker
Capacity (Metric) 250 mL
Capacity (English) 8.4 oz.
For Use With (Equipment) Ultrasonic Benchtop Cleaning Bath
Material Glass