Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beakers
Product Code. 15318644
Change view
Click to view available options
Capacity (English):
8.4 oz.
20.28 oz.
Capacity (Metric):
250 mL
600 mL
Material:
Glass
Stainless Steel
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Capacity (Metric) Material unitSize
15318644 20.28 oz. 600 mL Glass Each
10041654 20.28 oz. 600 mL Stainless Steel Each
15338644 8.4 oz. 250 mL Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15318644 Supplier Branson Ultrasonics™ Supplier No. 000140004
  • ONLINE EXCLUSIVE PRICE
    16.38€
    13.70€ / Pack of 1
    Save 2.68€
    16% Off
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
For use with Bransonic ultrasonic baths
  • Suitable for all models
  • Available in glass, stainless steel, or polypropylene
Specifications
Type Beaker
Capacity (Metric) 600 mL
Capacity (English) 20.28 oz.
For Use With (Equipment) Ultrasonic Benchtop Cleaning Bath
Material Glass