Learn More
BRANDTECH™ VITLAB™ POM or PMP Forceps
Product Code. 10377604
Click to view available options
Length (Metric):
180 mm
Tip Style:
Blunt
Material:
POM
Unit Size:
Pack of 5
Product Code. 10377604
Brand: BRANDTECH™ VITLAB™ 68299
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
POM or PMP forceps offer resilience and flexibility. Available in various lengths and tip styles.
POM (polyoxymethylene) forceps:
  • Color: yellow
  • External finger grooves for exceptional grip
  • Blunt tips
  • Part No. 68299 and 68399 have grooves inside of tip
  • Part No. 68099 and 68199 are without grooves
PMP (polymethylpentene) forceps:
  • Color: white
  • Pointed tips
Specifications
Tip Style Blunt
Length (English) 7 in.
Material POM
Length (Metric) 180 mm
Product Type Forceps
Quantity (No. per Case) 5/Pk.
Type POM Forceps
Style Standard