Learn More
VACUUBRAND™ Rotary Vane Vacuum Pump: Model RZ2.5
Product Code. 10635301
Change view
Click to view available options
Flow Rate:
1.4 to 1.7 CFM
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
10635301 1.4 to 1.7 CFM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10635301 Supplier VACUUBRAND™ Supplier No. 20698120
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed from the ground-up for use in laboratory applications
Designed for use where diaphragm pumps cannot provide sufficient vacuum depth. RZ2.5 has a free-air capacity of 47L/min. and a KF16 small flange inlet, with supplied 10mm (3/8 in.) polypropylene hose barb.
  • Dual -stage design for an ultimate vacuum of 1.5x10-3 Torr
  • Quiet, small and energy efficient
  • Rugged and low maintenance
For freeze dryers, benchtop centrifugal concentrators or other low-temperature evaporations of high boiling point solvents, molecular distillations, schlenk lines. As with all rotary vane pumps, a foreline cold trap is required for use in these applications.
TRUSTED_SUSTAINABILITY
Specifications
Horsepower 0.24 hp
Amperage 1.5/1.6 A
Includes Pump oil filled and completely mounted, ready for use, with manual.
Motor Speed 1500 to 1800
Certifications/Compliance UL
Electrical Requirements 230 V 50/60 Hz
Model RZ2.5
Noise Level 43 dB (A)
Plug Type CEE
Temperature (Metric) Storage -10°C to +60°C
Weight (English) 25.1 lb.
Weight (Metric) 11.4 kg
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 12°C
Flow Rate 1.4 to 1.7 CFM
For Use With Freeze dryers, benchtop centrifugal concentrators or other low-temperature evaporations of high boiling point solvents, molecular distillations
Frequency 50/60 Hz
Overall Height (English) 7.4 in.
Overall Height (Metric) 19 cm
Overall Length (English) 12.4 in.
Overall Length (Metric) 31.6 cm
Overall Width (English) 4.9 in.
Enclosure Rating IP40
Maximum Pressure 1.1 bar
Voltage 230 V