Learn More
VACUUBRAND™ B-oil for Rotary Vane Pumps
Product Code. p-4492948
Click to view available options
Capacity (Metric):
1 L
Unit Size:
1 L
Product Code. 10633952
Brand: VACUUBRAND™ 20687010
Edge
VACUUBRAND™ B-oil offers good chemical resistance, low vapor pressure, and improved stability when handling oxidants such as acidic or alkaline vapors. Suitable for initial filling of RE-, RZ-, RC- series of rotary vane pumps.
  • Low vapor pressure, even at elevated temperatures
  • Low oil backflow
  • Resistant to aging and breakdown
  • Minimum oxidation
  • Contains alkaline water to help prevent corrosion
  • Flashpoint: 260°C
  • Viscosity at 40°C: 94 mm2/sec
  • Density at 20°C: 0.87 g/cm
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1 L
For Use With Rotary vane vacuum pump
Maximum Pressure <1 x 10-3 mbar