Learn More
VACUUBRAND™ Oil-Free Chemistry Design Diaphragm Pump Key
Click to view available options
Overall Width (English):
2.59 in.
Unit Size:
Each
Description
- Diaphragm key (wrench) to replace diaphragms on VACUUBRAND 'NT' model vacuum pumps
Specifications
Specifications
| For Use With | Oil free diaphragm vacuum pump |
| Overall Width (English) | 2.59 in. |
| Type | Diaphragm Key |