Learn More
VACUUBRAND™ MD1C + AK + EK Oil-Free Chemistry Diaphragm Vacuum Pump
Product Code. 10603113
Click to view available options
Flow Rate:
25 L/min.
Unit Size:
Each
Product Code. 10603113
Brand: VACUUBRAND™ 20696620
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
An excellent choice for rotary evaporation and other laboratory applications
Suitable for benchtop rotary evaporators, and powerful enough for excellent results with high-boiling point solvents
  • Extremely low maintenance, no oil changes, no oil mist
  • Ultimate vacuum 1.5 Torr; free air capacity 25 liters per minute
  • Corrosion resistance
  • With an integrated gas ballast to provide consistent pump performance with condensed vapors
  • Adds solvent recovery, including an emissions condenser; compact three-stage design is suitable for benchtop rotary evaporation and gel drying with an SDS-PAGE concentration greater than 10%
Order Info
Included solvent recovery glassware for use in evaporative applications when no cold trap is used.
TRUSTED_SUSTAINABILITY
Specifications
Includes CEE plug
Electrical Requirements 230 V
Dimensions (L x W x H) 12.62 x 11.4 x 19 in. (32 x 29 x 48 cm)
Material PTFE (Pump Head)
Model MD1C+AK+EK
Weight (English) 22.48 lb.
Weight (Metric) 10.2 kg
Flow Rate 25 L/min.
Frequency 60 Hz
Overall Height (English) 19 in.
Overall Height (Metric) 48 cm
Overall Length (English) 12.62 in.
Overall Length (Metric) 32 cm
Overall Width (English) 11.4 in.
Overall Width (Metric) 29 cm
Voltage 230 V