Learn More
BRAND™ Transferpettor™ Pipet Tips
Product Code. 10673191 Shop All Brand Products
Click to view available options
Volume (Metric):
1 to 5 mL
100 to 500 μL
2 to 10 mL
200 to 1000 μL
Length (English):
6.14 in.
6.2 in.
Length (Metric):
15.6 cm
16 cm
Color:
Green
Orange
Red
Yellow
Unit Size:
Pack of 10
Product Code. 10673191
Brand: BRAND™ 702858
Only null left
Add to basket
Add to basket
Reusable Positive Displacement Pipet Tips
  • Made of Polyethylene
  • Reusable Positive Displacement Pipet Tips
  • Compatible to use with BrandTech Transferpettor Positive Displacement Pipettes
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Red
For Use With Transferpette/Transferpette S
Material Polypropylene
Volume (Metric) 1 to 5 mL
For Use With (Equipment) 1000 to 5000 μL Transferpettor
Length (English) 6.2 in.
Length (Metric) 16 cm