Learn More
BRAND™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Product Code. 10715883 Shop All Brand Products
Click to view available options
Volume (Metric):
0.1 to 1, 0.5 to 10, 10 to 100 μL
0.5 to 10, 10 to 100, 100 to 1000 μL
100 to 1000, 500 to 5000, 1000 to 10,000 μL
2 to 20, 20 to 200, 100 to 1000 μL
Includes:
D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
Type:
Macro Set
Micro Set
Pipette Set, Standard
Starter Set, Adjustable
Unit Size:
Each
Product Code. 10715883
Brand: BRAND™ 704792
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
Set contents:
  • Three adjustable volume pipettes
  • Three shelf/rack mounts
  • Three filled polypropylene Tip Boxes
Simple one-handed operation units exhibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
  • Available as Standard Set, Micro Set, Midsize Set and Macro Set
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With Laboratory
Number of Channels 1
Product Line Transferpette
Type Macro Set
Volume (Metric) 100 to 1000, 500 to 5000, 1000 to 10,000 μL
For Use With (Application) Demanding laboratory applications
Includes D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes