Learn More
BRAND™ White Real-Time PCR (qPCR) 96-Well Microplates, BIO-CERT™ PCR QUALITY, Standard Profile
GREENER_CHOICE
Product Code. 10696095 Shop All Brand Products
Click to view available options
Includes:
Half Skirt
No Skirt
Unit Size:
Pack of 50
Product Code. 10696095
Brand: BRAND™ 781357
Only null left
Add to basket
Add to basket
Designed optimized results when performing quantitative Real Time PCR (qPCR)
Designed for optimal performance in fluorescence signal analysis and DNA quantification, these plates are ideal for qPCR applications. Key features include:
  • Enhanced Signal Reflection: White surface and titanium dioxide (TiO2) coloring maximize fluorescence signal reflection for precise quantification.
  • Exceptional Purity: BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
  • Advanced Manufacturing Standards: Produced in state-of-the-art clean room conditions for consistent quality.
  • Superior Thermal Performance: Extra-thin walls provide efficient thermal transfer.
  • User-Friendly Design:
    • Smooth surfaces optimize signal reflection
    • Blue embossed alphanumeric coding and a cut corner design ensure easy orientation
    • Standard-profile, with an elevated rim offers compatibility with various thermal cyclers
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color White
Disposable Single-use
Product Type qPCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Semi-skirted
Material Polypropylene
Volume (Metric) Well 0.20 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.2 mL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Optimize results when performing quantitative RT-PCR
Includes Half Skirt
No. per Pack 50/Pk.
Profile Standard
RNase Free Yes