Learn More
BRAND™ BRANDplates™ Smooth-Walled Insert Strips
Shop All Brand Products
Click to view available options
Pore Size:
0.4 μm
1.0 μm
3.0 μm
8.0 μm
Item Description:
PC membrane
PET membrane
Unit Size:
Pack of 12
Description
Specifications
Specifications
| Pore Size | 0.4 μm |
| Type | Smooth-walled Insert Strip |
| Item Description | PC membrane |
| For Use With (Equipment) | 24-/6-well BRANDplates™ |
| Quantity | 12/Pk. |
| Surface Treatment | cellGrade plus |
| Sterility | Sterile |