Learn More
BRAND™ BRANDplates hydroGrade™ 96-Well Microplates
Product Code. 10012403 Shop All Brand Products
Click to view available options
Color:
Clear
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Total Volume (Metric) Well:
330 μL
350 μL
Unit Size:
Pack of 100
Product Code. 10012403
Brand: BRAND™ 781780
Featuring a strongly hydrophilic surface for binding of hydrophilic molecules or for homogeneous assays with hydrophobic molecules that remain in solution
  • Immunoassay surface with strong affinity to hydrophilic molecules, such as glycoproteins and peptides, antibodies with predominantly hydrophilic regions, and nucleic acids
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Binding Property Hydrophilic
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment hydroGrade
Total Volume (Metric) Well 330 μL
Well Shape Round Bottom (U-well)
For Use With (Application) Immobilization of hydrophilic molecules
Quantity 100/Cs.