Learn More
Brady™ BradyPrinter i7100 - EU with Brady Workstation LAB Suite
Product Code. 17137021
Click to view available options
Resolution:
300 dpi
600 dpi
Description:
With Software, Peel Function
With Softwate
Unit Size:
1 set
Product Code. 17137021
Brand: Brady™ 306889
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the Aerospace, Defence and Mass Transit, Electronics, Automotive, Logistics, Electrical, Voice-Datacom and LAB markets.
  • Industrial thermal transfer benchtop printer
  • Ideal for customers requiring reliable high volume printing on lots of different materials within the following markets:
    • Aerospace
    • Defence and Mass Transit
    • Electronics
    • Automotive
    • Logistics
    • Electrical
    • Voice-Datacom
    • LAB
  • It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more
  • The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels
  • This suite combines frequently used software apps to create laboratory identification labels
Specifications
Product Type Industrial Printer
Display Type Color Touchscreen
Print Technology Thermal Transfer
Width (Metric) 252.00 mm
Height (Metric) 288.00 mm
Depth (Metric) 460.00 mm
Dimensions (L x W x H) 460.00 x 252.00 x 288.00 mm
Color Capability Monochrome
Description With Software, Peel Function
Includes Peel and Present module pre-installed. It ships with an EU power cord, 2 empty ribbon cores, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English, a printed user manual in English and Brady Workstation Laboratory ID Suite
Print Speed 150.00 mm/s
Resolution 600 dpi
Software Type Brady Workstation LAB Suite
Power Supply Details AC Adaptor
Alternate Part Number i7100-600P-EU-LABS