Learn More
Brady™ Printer i5100 - UK/EU with Brady Workstation LAB Suite
Product Code. 17147011
Click to view available options
Resolution:
300 dpi
600 dpi
Description:
With Cutter and Software
With Software
Unit Size:
1 set
Product Code. 17147011
Brand: Brady™ 306874
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The Brady Printer i5100 Industrial Label Printer provides the fundamental printing capabilities for the high-mix and high-volume work you need to get done every day.
  • The BradyPrinter i5100 Industrial Label Printer offers a perfect mix of performance & smarts
  • This benchtop printer can print on 70 label materials, more than 1200 stock label parts and tailored custom label materials and parts
  • Applications include a wide range of specialised and commonly used product labels, electronics and PCB labels, logistics and asset tracking labels, cable & component identification labels and laboratory sample labels
  • The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels
  • This suite combines frequently used software apps to create laboratory identification labels.
Specifications
Display Type Color Touchscreen
Print Technology Thermal Transfer
Width (Metric) 240.00 mm
Height (Metric) 317.00 mm
Depth (Metric) 435.00 mm
Dimensions (L x W x H) 435.00 x 240.00 x 317.00 mm
Color Capability Monochrome
Description With Cutter and Software
Includes Power cord, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English and a printed user manual in English, an RFID enabled roll holder, a ribbon take up spindle and a Brady Workstation Laboratory Identification Suite
Print Speed 300.00 mm/s
Resolution 300 dpi
Power Supply Details AC Adaptor