Learn More
Bradykinin (1-5), Thermo Scientific Chemicals
Product Code. p-5384580
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Product Code. 15475489
Brand: Thermo Scientific Alfa Aesar J63233.#0
Request Bulk or Custom Format
For cellular and molecular biology applications
Biologically active fragment of bradykinin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) Biologically active fragment of bradykinin
Molecular Weight (g/mol) 572.67
Product Type Bradykinin (1-5)
Quantity 5 mg
Regulatory Status RUO - research use only
Sequence H-Arg-Pro-Pro-Gly-Phe-OH