Learn More
Brady™ BradyJet™ Pre-Cut Polyester Labels
Product Code. 16100491
Click to view available options
Dimensions (LxW):
3.35 x 2.17 in.
4.33 x 2.95 in.
6.10 x 4.13 in.
Unit Size:
Each
Product Code. 16100491
Brand: Brady™ 150026
Only null left
Add to basket
Add to basket
Brady™ BradyJet™ Pre-Cut Polyester Labels are for use with BradyJet™ J2000 and are ideal for a wide range of identification applications.
  • Constructed of indoor-use general purpose polyester
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Roll
Material Polyester
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking, Lockout Tagout Labeling, GHS and Chemical Labels, Arc Flash Labels
For Use With (Equipment) BradyJet™ J2000
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 340 Labels/Bag
Quantity 340/Roll
Shape Rectangle
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (LxW) 3.35 x 2.17 in.
Length (English) 2.17 in.
Width (English) 2.17 in.
Width (Metric) 55.12 mm
Height (English) 3.35 in.
Height (Metric) 85.09 mm
Diameter (English) 1.88 in. I.D.
Alternate Part Number J20-260-2569