Learn More
Brady™ Brady Workstation Safety and Facility Identification Software Suite, E-Media
Click to view available options
:
Brady Workstation Safety and Facility Identification Software Suite, E-Media
Description
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Features:
- Custom Designer - Complete, customized label creation and editing.
- Express Signs - Create safety and facility signs in minutes with predefined templates and layouts.
- Arc Flash - Create NFPA compliant arc flash labels in a few clicks.
- Arrow Tape - Create arrow tape labels with predefined templates to show pipe flow direction.
- Pipemarker - Easily create ANSI/ASME compliant pipe markers using pre-defined templates.
- Sequence - Easily create labels based on numeric and alphanumeric sequences.
Specifications
Specifications
| Product Type | Software |
| For Use With (Equipment) | Brady Label Printers |
| Width (English) | 0.002 in. |
| Format | Email with Activation Code |
| Includes | Email with activation code |
| Languages | Bulgarian;Chinese (Simplified);Chinese (Traditional);Croatian;Czech;Danish;Dutch;English (US);Estonian;Finnish;French;German;Hungarian;Italian;Japanese;Korean;Norwegian (Bokmål);Polish;Portuguese;Romanian;Russian;Slovak;Slovenian;Spanish (Mexico);Spanish;Swedish;Turkish;Vietnamese |
| Resolution | 1024 x 768 |
| Software Type | Brady Workstation Safety and Facility Identification Software Suite |
| Operating System | Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10 |
| Alternate Part Number | BWS-SFIDS-NA-EM |