Learn More
Brady™ TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels
Product Code. 11532905
Click to view available options
Quantity:
100/Roll
500/Pk.
Height (Metric):
12.7 mm
25 mm
55.88 mm
66.675 mm
Material:
Nylon Cloth
Polyester
Vinyl
Unit Size:
1 roll
Product Code. 11532905
Brand: Brady™ 062259
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels are ideal for cables, laboratory ID or general labeling where graphics need to be protected.
  • Labels for the TLS 2200™ and TLS-PC Link™ thermal transfer Portable Printers.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive , good print smudge resistance, solvent resistance and high/low temperature performance.
Specifications
Certifications/Compliance RoHS
Description TLS 2200/TLS PC Link Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
Material Vinyl
Product Type TLS 2200™/TLS-PC Link™ Label
For Use With (Application) Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube
For Use With (Equipment) Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube
Labels per Row 1
Quantity 100/Roll
Shape Rectangle
Temperature Range (English) -40 to 158°F
Temperature Range (Metric) -40°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 60.33 mm
Wire Gauge 3 to 000 AWG
Width (English) 2.2 in.
Width (Metric) 12.70 mm
Height (English) 0.5 in.
Height (Metric) 55.88 mm