Learn More
Brady™ TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels
Product Code. 10512212
Click to view available options
Quantity:
100/Roll
500/Pk.
Height (Metric):
12.7 mm
25 mm
55.88 mm
66.675 mm
Material:
Nylon Cloth
Polyester
Vinyl
Unit Size:
1 roll
Product Code. 10512212
Brand: Brady™ 018506
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels are ideal for cables, laboratory ID or general labeling where graphics need to be protected.
  • Labels for the TLS 2200™ and TLS-PC Link™ thermal transfer Portable Printers.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive , good print smudge resistance, solvent resistance and high/low temperature performance.
Specifications
Certifications/Compliance RoHS
Description TLS 2200/TLS PC Link Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
Material Nylon Cloth
Product Type TLS 2200™/TLS-PC Link™ Label
Sterility Non-sterile
For Use With (Application) Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube
For Use With (Equipment) BMP™61 Labeling System; BMP™71 Label Printer; TLS 2200™ Thermal Labeling System;TLS-PC LINK™ Thermal Labeling System
Labels per Row 1
Quantity 500/Pk.
Shape Rectangle
Temperature Range (English) 193 to 293°F
Temperature Range (Metric) 90 to 145°C
Tensile Strength 80 lb./in.
Thickness (English) 0.0065 in.
Thickness (Metric) 0.165 mm
Vertical Repeat (Metric) 28.57 mm
Wire Gauge 3 AWG
Dimensions (LxW) 1 in. H x 0.375 in. W
Width (English) 0.3 in.
Width (Metric) 9.53 mm
Height (English) 1 in.
Height (Metric) 25 mm