Learn More
Brady™ and TLS-PC Link™ Tamper-Evident Labels
Product Code. 10542402
Click to view available options
Color:
White
Unit Size:
1 roll
Product Code. 10542402
Brand: Brady™ 018700
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
and TLS-PC Link™ Labels are made of Tamper-Resistant Vinyl (B-351) that is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
  • High performance white matte vinyl with permanent adhesive is ideal for applications that require resistance to product tampering
  • Use caution when removing from liner as material is fragile
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -40 to 180° F (-40 to 82° C)
Specifications
Certifications/Compliance RoHS, 2005/618/EC MCV, 2002/95/EC
Description Surface Mount
Adhesive Type Acrylic
Color White
Finish Matte
Format Label
Material Vinyl
Product Type TLS 2200™/TLS-PC Link™ Label
Sterility Non-sterile
For Use With (Application) For rating and serial plates that require high performance and resistance to product tampering
For Use With (Equipment) BMP™71, TLS 2200™, TLS-PC Link™
Labels per Row 1
Quantity 100/Roll
Shape Rectangle
Temperature Range (Metric) -40°C to +80°C
Tensile Strength 63 lb./in.
Thickness (English) 0.0028 in.
Thickness (Metric) 0.066 mm
Vertical Repeat (Metric) 28.57 mm
Dimensions (LxW) 1 in. H x 2 in. W
Width (English) 2 in.
Width (Metric) 50.8 mm
Height (English) 1 in.
Height (Metric) 25.4 mm