Learn More
Brady™ Thermal Transfer Printable Labels
Product Code. 12069857
Click to view available options
Width (English):
1.625 in.
Height (English):
0.6 in.
Quantity:
3000/Pk.
Material:
Polyester
Unit Size:
1 roll
Product Code. 12069857
Brand: Brady™ 062365
Nylon cloth labels for thermal transfer printers
  • Feature StainerBondz™ super chemical resistant polyester
  • B-481 material, coupled with R6410 series ribbon, will remain resistant when exposed to xylene and other chemicals that are commonly used in the slide staining process
  • No print removal, no label buckling, and no discoloration that would cause bar codes to become unreadable
  • Permanent acrylic adhesive
  • Label stays adhered to the slide
Specifications
Description Thermal Transfer Printable Labels
Adhesive Type Permanent Acrylic
Color White
Display Consignment
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Sterility Non-sterile
For Use With (Application) General Identification, Slide label ID
For Use With (Equipment) BBP™ 81, Brady 1244, Brady 1344, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, IP300, IP600, Tagus T200, Tagus T300, Thermal Transfer Printers
Labels per Row 1
Quantity 3000/Pk.
Shape Rectangle
Temperature Range (Metric) -196°C to +110°C
Thickness (Metric) 0.048 mm
Vertical Repeat (Metric) 19.05 mm
Dimensions (LxW) 0.6 x 1.625 in.
Length (English) 0.6 in.
Width (English) 1.625 in.
Width (Metric) 15.24 mm
Height (English) 0.6 in.
Height (Metric) 15.24 mm
Height (Metric) Printable Area 25.40 mm