Learn More
Brady™ Bradylink™ Connecting Links
Product Code. 11731296 Shop All Brady Products
Change view
Click to view available options
Color:
Yellow
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11731296 Yellow 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11731296 Supplier Brady™ Supplier No. 078246
  • ONLINE EXCLUSIVE PRICE
    67.15€
    66.02€ / Pack of 1
    Save 1.13€
    1% Off
Estimated Shipment: 08-01-2027
Used to permanently attach lengths of chain or to repair chain link
  • For indoor use only
  • Link design prevents kinking
  • Connecting Links match our posts and chain
Specifications
Length (English) 2 in.
Length (Metric) 5.08 cm
Packaging Quantity 36/Pk.
Width (Metric) 50.00 mm
Color Yellow
Material B-900 Polyethylene
Type Connecting Links