Learn More
Brady™ BBP™ WARNING Pre-Printed Pre-Cut Blank Sign Headers
Product Code. 11727254
Change view
Click to view available options
Dimensions (L x W):
4 x 6 in.
57 x 75 mm
Quantity:
175/Pk.
300/Pk.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity unitSize
11727254 4 x 6 in. 175/Pk. Each
11727324 57 x 75 mm 300/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11727254 Supplier Brady™ Supplier No. 142186
  • ONLINE EXCLUSIVE PRICE
    489.00€
    415.23€ / Pack of 1
    Save 73.77€
    15% Off
In Stock
Add to basket
Add to basket
BBP™ WARNING Pre-Printed Pre-Cut Blank Sign Headers are durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
Specifications
Certifications/Compliance ASME ANSI Z535, OSHA 1910.145
Alternate Part Number B30-25-595-ANSIWA
Finish Gloss
For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Material Vinyl
Quantity 175/Pk.
Adhesive Type Permanent Acrylic
Description Pre-Printed Pre-Cut Blank Sign headers for BBP3x Printers
For Use With (Application) Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
Height (English) 4 in.
Height (Metric) 152 mm
Max. Temperature (English) 180°F
Min. Temperature (English) -40°F
Thickness 0.004 in.
Width (English) 6 in.
Width (Metric) 152.4 mm
Temperature Range (Metric) -40°C to 82°C
Temperature Range (English) -40°F to 180 °F
Languages English
Dimensions (L x W) 4 x 6 in.
Displayed Text Warning
Product Type Indoor/Outdoor Tape
Color Black on Orange